Assay Details
| Gene Symbol: Nme6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018757.1 | NP_061227.1 | 4 - 5 | 361 | 64 | |
| GenBank mRNA | AF051942.1 | - | 4 - 5 | 361 | 64 | |
| AK004201.1 | - | 4 - 5 | 271 | 64 | ||
| BC002007.1 | - | 4 - 5 | 361 | 3593804 | 64 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nme6
Genomic Map