Assay Details
| Gene Symbol: Asxl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039939.1 | NP_001035028.1 | 10 - 11 | 1177 | 65 | |
| GenBank mRNA | AK122413.1 | - | 10 - 11 | 1177 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Asxl1
Genomic Map