Assay Details
| Gene Symbol: Gypa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010369.3 | NP_034499.3 | 4 - 5 | 297 | 65 | |
| GenBank mRNA | AK145998.1 | - | 4 - 5 | 298 | 65 | |
| AK157851.1 | - | 4 - 5 | 298 | 65 | ||
| AK162614.1 | - | 4 - 5 | 282 | 65 | ||
| BC148192.1 | - | 4 - 5 | 561 | 40045496 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gypa
Genomic Map