Assay Details
| Gene Symbol: Adgre5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163029.1 | NP_001156501.1 | 16 - 17 | 2224 | 57 | |
| NM_001163030.1 | NP_001156502.1 | 17 - 18 | 2371 | 57 | ||
| NM_001163031.1 | NP_001156503.1 | 16 - 17 | 2218 | 57 | ||
| NM_011925.2 | NP_036055.2 | 18 - 19 | 2506 | 57 | ||
| GenBank mRNA | AF146344.1 | - | 16 - 17 | 2095 | 57 | |
| AK004577.1 | - | 16 - 17 | 2234 | 57 | ||
| AK008101.1 | - | 4 - 5 | 515 | 57 | ||
| BC006676.1 | - | 16 - 17 | 2086 | 3499451 | 57 | |
| CT010326.1 | - | 16 - 17 | 2050 | 57 | ||
| Y18365.1 | - | 18 - 19 | 2364 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adgre5
Genomic Map