Assay Details
| Gene Symbol: Adgre5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163029.1 | NP_001156501.1 | 2 - 3 | 90 | ||
| NM_001163030.1 | NP_001156502.1 | 2 - 3 | 90 | |||
| NM_011925.2 | NP_036055.2 | 2 - 3 | 90 | |||
| GenBank mRNA | AF146344.1 | - | 2 - 3 | 90 | ||
| BB872151.1 | - | 2 - 3 | 90 | |||
| BC006676.1 | - | 2 - 3 | 3499451 | 90 | ||
| CD553351.1 | - | 2 - 3 | 90 | |||
| CT010326.1 | - | 2 - 3 | 90 | |||
| Y18365.1 | - | 2 - 3 | 124 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adgre5
Genomic Map