Assay Details
| Gene Symbol: Clgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009904.2 | NP_034034.2 | 11 - 12 | 1584 | 75 | |
| GenBank mRNA | AK019534.1 | - | 11 - 12 | 1498 | 75 | |
| AK034525.1 | - | 11 - 12 | 1485 | 75 | ||
| BC050767.1 | - | 11 - 12 | 1584 | 6771060 | 75 | |
| D14117.1 | - | 11 - 12 | 1597 | 75 | ||
| D86323.1 | - | 11 - 12 | 1597 | 75 | ||
More Information
Set Membership: |
|

Clgn
Genomic Map