Assay Details
| Gene Symbol: Clgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009904.2 | NP_034034.2 | 4 - 5 | 494 | 99 | |
| GenBank mRNA | AK019534.1 | - | 4 - 5 | 408 | 99 | |
| AK034525.1 | - | 4 - 5 | 396 | 99 | ||
| BC050767.1 | - | 4 - 5 | 494 | 6771060 | 99 | |
| D14117.1 | - | 4 - 5 | 507 | 99 | ||
| D86323.1 | - | 4 - 5 | 507 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Clgn
Genomic Map