| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800019406.mm.1 |
NM_027179.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019406.mm.1 |
NM_027179.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019406.mm.1 |
NM_134118.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019406.mm.1 |
NM_134118.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019406.mm.1 |
TC0800002553.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800019402.mm.1 |
NM_027179.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019402.mm.1 |
NM_027179.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019402.mm.1 |
NM_134118.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019402.mm.1 |
NM_134118.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019402.mm.1 |
TC0800002553.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800019407.mm.1 |
NM_027179.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019407.mm.1 |
NM_027179.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019407.mm.1 |
NM_134118.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019407.mm.1 |
NM_134118.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019407.mm.1 |
TC0800002553.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800019404.mm.1 |
NM_027179.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019404.mm.1 |
NM_027179.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019404.mm.1 |
NM_134118.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019404.mm.1 |
NM_134118.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019404.mm.1 |
TC0800002553.mm.1 |
MTA TC |