Assay Details
| Gene Symbol: Ddx39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_197982.3 | NP_932099.2 | 1 - 2 | 159 | 105 | |
| GenBank mRNA | AK012002.1 | - | 1 - 2 | 69 | 105 | |
| AK016960.1 | - | 1 - 2 | 69 | 105 | ||
| AK137114.1 | - | 65 | 105 | |||
| AK145927.1 | - | 1 - 2 | 102 | 105 | ||
| AK146294.1 | - | 1 - 2 | 104 | 105 | ||
| AK149475.1 | - | 1 - 2 | 70 | 105 | ||
| AK150997.1 | - | 1 - 2 | 88 | 105 | ||
| AK151395.1 | - | 1 - 2 | 69 | 105 | ||
| AK152394.1 | - | 1 - 2 | 69 | 105 | ||
| AK162752.1 | - | 627 | 105 | |||
| AK172448.1 | - | 1 - 2 | 88 | 105 | ||
| BC020134.1 | - | 1 - 2 | 47 | 3969048 | 105 | |
| BY065389.1 | - | 1 - 2 | 138 | 105 | ||
| CF172497.1 | - | 1 - 2 | 56 | 105 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK137114.1, AK145927.1, AK146294.1, AK150997.1, AK151395.1, AK152394.1, AK162752.1, AK172448.1, BC020134.1, NM_197982.3 transcripts.
Set Membership: |
|

Ddx39
Genomic Map