Assay Details
| Gene Symbol: Ddx39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_197982.3 | NP_932099.2 | 3 - 4 | 500 | 59 | |
| GenBank mRNA | AK011285.1 | - | 2 - 3 | 340 | 59 | |
| AK012002.1 | - | 3 - 4 | 410 | 59 | ||
| AK016960.1 | - | 3 - 4 | 410 | 59 | ||
| AK137114.1 | - | 406 | 59 | |||
| AK145927.1 | - | 3 - 4 | 443 | 59 | ||
| AK146294.1 | - | 3 - 4 | 445 | 59 | ||
| AK149475.1 | - | 3 - 4 | 406 | 59 | ||
| AK150997.1 | - | 3 - 4 | 429 | 59 | ||
| AK151395.1 | - | 3 - 4 | 410 | 59 | ||
| AK152394.1 | - | 3 - 4 | 410 | 59 | ||
| AK162752.1 | - | 968 | 59 | |||
| AK164289.1 | - | 3 - 4 | 370 | 59 | ||
| AK167890.1 | - | 3 - 4 | 370 | 59 | ||
| AK168079.1 | - | 3 - 4 | 369 | 59 | ||
| AK172448.1 | - | 3 - 4 | 429 | 59 | ||
| BC020134.1 | - | 3 - 4 | 388 | 3969048 | 59 | |
| CF172497.1 | - | 3 - 4 | 397 | 59 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK151395.1 transcripts.
Set Membership: |
|

Ddx39
Genomic Map