Assay Details
| Gene Symbol: Ddx39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_197982.3 | NP_932099.2 | 5 - 6 | 785 | 67 | |
| GenBank mRNA | AK011285.1 | - | 4 - 5 | 624 | 67 | |
| AK137114.1 | - | 691 | 67 | |||
| AK145927.1 | - | 5 - 6 | 728 | 67 | ||
| AK146294.1 | - | 5 - 6 | 730 | 67 | ||
| AK150997.1 | - | 5 - 6 | 714 | 67 | ||
| AK152394.1 | - | 5 - 6 | 695 | 67 | ||
| AK162752.1 | - | 1253 | 67 | |||
| AK164289.1 | - | 5 - 6 | 655 | 67 | ||
| AK167890.1 | - | 5 - 6 | 655 | 67 | ||
| AK168079.1 | - | 5 - 6 | 654 | 67 | ||
| AK172448.1 | - | 5 - 6 | 714 | 67 | ||
| BC020134.1 | - | 5 - 6 | 673 | 3969048 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx39
Genomic Map