Assay Details
| Gene Symbol: Cdc27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285988.1 | NP_001272917.1 | 17 - 18 | 2487 | 144 | |
| NM_001285989.1 | NP_001272918.1 | 17 - 18 | 2469 | 144 | ||
| NM_001285990.1 | NP_001272919.1 | 16 - 17 | 2392 | 144 | ||
| NM_145436.2 | NP_663411.2 | 17 - 18 | 2472 | 144 | ||
| GenBank mRNA | AK148278.1 | - | 17 - 18 | 2328 | 144 | |
| AK161143.1 | - | 6 - 7 | 848 | 144 | ||
| AK161471.1 | - | 17 - 18 | 2337 | 144 | ||
| BC157955.1 | - | 17 - 18 | 2486 | 9088128 | 144 | |
| BC172100.1 | - | 17 - 18 | 2486 | 9054386 | 144 | |
| BC172149.1 | - | 16 - 17 | 2391 | 144 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdc27
Genomic Map