Assay Details
| Gene Symbol: Crhr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007762.4 | NP_031788.1 | 10 - 11 | 1120 | 58 | |
| GenBank mRNA | AB375515.1 | - | 10 - 11 | 930 | 58 | |
| AF483484.1 | - | 10 - 11 | 930 | 58 | ||
| AF483485.1 | - | 10 - 11 | 930 | 58 | ||
| AK144430.1 | - | 10 - 11 | 1188 | 58 | ||
| AK158806.1 | - | 10 - 11 | 1121 | 58 | ||
| BC103675.1 | - | 10 - 11 | 948 | 40046992 | 58 | |
| BC104735.1 | - | 11 - 12 | 948 | 40046995 | 58 | |
| BC105673.1 | - | 11 - 12 | 1024 | 40046994 | 58 | |
| BC105675.1 | - | 10 - 11 | 948 | 40046993 | 58 | |
| FJ668671.1 | - | 9 - 10 | 810 | 58 | ||
| X72305.1 | - | 10 - 11 | 961 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Crhr1
Genomic Map