Assay Details
| Gene Symbol: Crhr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007762.4 | NP_031788.1 | 7 - 8 | 901 | 64 | |
| GenBank mRNA | AB375515.1 | - | 7 - 8 | 711 | 64 | |
| AF483484.1 | - | 7 - 8 | 711 | 64 | ||
| AF483485.1 | - | 7 - 8 | 711 | 64 | ||
| AK144430.1 | - | 7 - 8 | 969 | 64 | ||
| AK158806.1 | - | 7 - 8 | 902 | 64 | ||
| BC103675.1 | - | 7 - 8 | 729 | 40046992 | 64 | |
| BC104735.1 | - | 8 - 9 | 729 | 40046995 | 64 | |
| BC105673.1 | - | 8 - 9 | 805 | 40046994 | 64 | |
| BC105675.1 | - | 7 - 8 | 729 | 40046993 | 64 | |
| FJ668671.1 | - | 6 - 7 | 591 | 64 | ||
| X72305.1 | - | 7 - 8 | 742 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Crhr1
Genomic Map