Assay Details
| Gene Symbol: Timm22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291161.1 | NP_001278090.1 | 2 - 3 | 421 | 72 | |
| NM_019818.5 | NP_062792.2 | 2 - 3 | 502 | 72 | ||
| NR_111896.1 | 2 - 3 | 410 | 72 | |||
| GenBank mRNA | AF223950.1 | - | 2 - 3 | 468 | 72 | |
| AK011745.1 | - | 2 - 3 | 454 | 72 | ||
| AK012130.2 | - | 2 - 3 | 456 | 72 | ||
| AK017389.1 | - | 2 - 3 | 455 | 72 | ||
| AK028132.1 | - | 2 - 3 | 454 | 72 | ||
| AK153837.1 | - | 2 - 3 | 457 | 72 | ||
| BC022610.1 | - | 2 - 3 | 438 | 72 | ||
| BC055279.1 | - | 2 - 3 | 466 | 5688754 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Timm22
Genomic Map