Assay Details
| Gene Symbol: Pitpna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008850.1 | NP_032876.1 | 2 - 3 | 278 | 89 | |
| GenBank mRNA | AK144618.1 | - | 2 - 3 | 291 | 89 | |
| AK149745.1 | - | 2 - 3 | 268 | 89 | ||
| AK151496.1 | - | 2 - 3 | 267 | 89 | ||
| AK152089.1 | - | 2 - 3 | 291 | 89 | ||
| AK152874.1 | - | 2 - 3 | 284 | 89 | ||
| AK153569.1 | - | 2 - 3 | 291 | 89 | ||
| AK165659.1 | - | 2 - 3 | 283 | 89 | ||
| AK167760.1 | - | 2 - 3 | 291 | 89 | ||
| AK168187.1 | - | 2 - 3 | 248 | 89 | ||
| AK168720.1 | - | 2 - 3 | 281 | 89 | ||
| AK187087.1 | - | 2 - 3 | 91 | 89 | ||
| AK187515.1 | - | 2 - 3 | 64 | 89 | ||
| BC056171.1 | - | 2 - 3 | 233 | 5710926 | 89 | |
| S72681.1 | - | 2 - 3 | 53 | 89 | ||
| U96725.1 | - | 2 - 3 | 279 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pitpna
Genomic Map