Assay Details
| Gene Symbol: Ift52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172150.4 | NP_742162.2 | 2 - 3 | 187 | 117 | |
| GenBank mRNA | AK088467.1 | - | 2 - 3 | 188 | 117 | |
| AK144188.1 | - | 2 - 3 | 287 | 117 | ||
| AK160745.1 | - | 2 - 3 | 192 | 117 | ||
| AK167194.1 | - | 2 - 3 | 203 | 117 | ||
| BC037708.1 | - | 2 - 3 | 156 | 4222723 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ift52
Genomic Map