| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100017636.mm.1 |
NM_170758.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100017636.mm.1 |
NM_170758.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100017636.mm.1 |
TC1100001882.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100017640.mm.1 |
NM_170758.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100017640.mm.1 |
NM_170758.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100017640.mm.1 |
TC1100001882.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100017641.mm.1 |
NM_170758.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100017641.mm.1 |
NM_170758.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100017641.mm.1 |
TC1100001882.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100017637.mm.1 |
NM_170758.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100017637.mm.1 |
NM_170758.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100017637.mm.1 |
TC1100001882.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100017643.mm.1 |
NM_170758.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100017643.mm.1 |
NM_170758.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100017643.mm.1 |
TC1100001882.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100017638.mm.1 |
NM_170758.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100017638.mm.1 |
NM_170758.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100017638.mm.1 |
TC1100001882.mm.1 |
MTA TC |