Assay Details
| Gene Symbol: Cd300a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170758.3 | NP_739564.1 | 4 - 5 | 971 | 100 | |
| GenBank mRNA | AB065156.1 | - | 4 - 5 | 690 | 100 | |
| AB091765.1 | - | 4 - 5 | 697 | 100 | ||
| AB091766.1 | - | 4 - 5 | 693 | 100 | ||
| AB095675.1 | - | 4 - 5 | 794 | 100 | ||
| AK045869.1 | - | 4 - 5 | 971 | 100 | ||
| AK053160.1 | - | 4 - 5 | 647 | 100 | ||
| BC139296.1 | - | 4 - 5 | 907 | 8862318 | 100 | |
| BC139297.1 | - | 4 - 5 | 907 | 8862319 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cd300a
Genomic Map