Assay Details
| Gene Symbol: Prdm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK028414.1 | - | 1 - 2 | 126 | 126 | |
| CB247385.1 | - | 1 - 2 | 143 | 126 | ||
| CD350319.1 | - | 1 - 2 | 184 | 126 | ||
More Information
Set Membership: |
|

Prdm4
Genomic Map