Assay Details
| Gene Symbol: Zdhhc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133967.3 | NP_598728.1 | 1 - 2 | 385 | 100 | |
| GenBank mRNA | AK041905.1 | - | 1 - 2 | 254 | 100 | |
| AK089681.1 | - | 229 | 100 | |||
| AK090333.1 | - | 1 - 2 | 229 | 100 | ||
| AK157968.1 | - | 1 - 2 | 238 | 100 | ||
| AK170639.1 | - | 1 - 2 | 238 | 100 | ||
| AK170923.1 | - | 1 - 2 | 208 | 100 | ||
| AK172462.1 | - | 1 - 2 | 219 | 100 | ||
| BC075666.1 | - | 1 - 2 | 385 | 30605163 | 100 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK041905.1, AK157968.1, AK170639.1, AK170923.1, AK172462.1, BC075666.1, NM_133967.3 transcripts.
Set Membership: |
|

Zdhhc7
Genomic Map