Assay Details
| Gene Symbol: Zdhhc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133967.3 | NP_598728.1 | 3 - 4 | 798 | 56 | |
| GenBank mRNA | AK041905.1 | - | 3 - 4 | 667 | 56 | |
| AK089681.1 | - | 643 | 56 | |||
| AK090333.1 | - | 3 - 4 | 643 | 56 | ||
| AK157968.1 | - | 3 - 4 | 651 | 56 | ||
| AK170639.1 | - | 3 - 4 | 651 | 56 | ||
| AK170923.1 | - | 3 - 4 | 621 | 56 | ||
| AK172462.1 | - | 3 - 4 | 632 | 56 | ||
| AK201914.1 | - | 1 - 2 | 156 | 56 | ||
| AY166673.1 | - | 1 - 2 | 316 | 56 | ||
| BC013467.1 | - | 2 - 3 | 349 | 4218025 | 56 | |
| BC075666.1 | - | 3 - 4 | 798 | 30605163 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zdhhc7
Genomic Map