Assay Details
| Gene Symbol: Zdhhc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133967.3 | NP_598728.1 | 5 - 6 | 1018 | 92 | |
| GenBank mRNA | AK041905.1 | - | 5 - 6 | 887 | 92 | |
| AK089681.1 | - | 863 | 92 | |||
| AK090333.1 | - | 5 - 6 | 864 | 92 | ||
| AK157968.1 | - | 5 - 6 | 871 | 92 | ||
| AK170639.1 | - | 5 - 6 | 871 | 92 | ||
| AK170923.1 | - | 5 - 6 | 841 | 92 | ||
| AK172462.1 | - | 5 - 6 | 852 | 92 | ||
| AK213858.1 | - | 2 - 3 | 146 | 92 | ||
| AY166673.1 | - | 3 - 4 | 536 | 92 | ||
| BC013467.1 | - | 4 - 5 | 569 | 4218025 | 92 | |
| BC075666.1 | - | 5 - 6 | 1018 | 30605163 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zdhhc7
Genomic Map