Assay Details
| Gene Symbol: Usp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009462.1 | NP_033488.1 | 12 - 13 | 2195 | 91 | |
| GenBank mRNA | AK077453.1 | - | 12 - 13 | 2217 | 91 | |
| AK129083.1 | - | 12 - 13 | 2203 | 91 | ||
| AK153675.1 | - | 12 - 13 | 2687 | 91 | ||
| AK157970.1 | - | 12 - 13 | 2219 | 91 | ||
| AK165156.1 | - | 12 - 13 | 2215 | 91 | ||
| AK166243.1 | - | 12 - 13 | 2211 | 91 | ||
| AK172443.1 | - | 12 - 13 | 2217 | 91 | ||
| BC007134.1 | - | 12 - 13 | 2197 | 3491297 | 91 | |
| D84096.1 | - | 12 - 13 | 2195 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Usp10
Genomic Map