Assay Details
| Gene Symbol: Usp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009462.1 | NP_033488.1 | 2 - 3 | 151 | 111 | |
| GenBank mRNA | AK077453.1 | - | 2 - 3 | 175 | 111 | |
| AK129083.1 | - | 2 - 3 | 162 | 111 | ||
| AK137015.1 | - | 2 - 3 | 173 | 111 | ||
| AK153675.1 | - | 2 - 3 | 643 | 111 | ||
| AK157970.1 | - | 2 - 3 | 175 | 111 | ||
| AK165156.1 | - | 2 - 3 | 174 | 111 | ||
| AK172443.1 | - | 2 - 3 | 173 | 111 | ||
| BC007134.1 | - | 2 - 3 | 156 | 3491297 | 111 | |
| D84096.1 | - | 2 - 3 | 151 | 111 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Usp10
Genomic Map