Assay Details
| Gene Symbol: Usp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009462.1 | NP_033488.1 | 5 - 6 | 1335 | 67 | |
| GenBank mRNA | AK077453.1 | - | 5 - 6 | 1357 | 67 | |
| AK129083.1 | - | 5 - 6 | 1343 | 67 | ||
| AK137015.1 | - | 5 - 6 | 1357 | 67 | ||
| AK153675.1 | - | 5 - 6 | 1827 | 67 | ||
| AK157970.1 | - | 5 - 6 | 1359 | 67 | ||
| AK165156.1 | - | 5 - 6 | 1355 | 67 | ||
| AK166243.1 | - | 5 - 6 | 1352 | 67 | ||
| AK172443.1 | - | 5 - 6 | 1357 | 67 | ||
| BC007134.1 | - | 5 - 6 | 1337 | 3491297 | 67 | |
| D84096.1 | - | 5 - 6 | 1335 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Usp10
Genomic Map