Assay Details
| Gene Symbol: Mbtps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167910.1 | NP_001161382.1 | 21 - 22 | 3271 | 62 | |
| NM_019709.4 | NP_062683.3 | 22 - 23 | 3420 | 62 | ||
| GenBank mRNA | AF094820.1 | - | 20 - 21 | 2839 | 62 | |
| AK029048.1 | - | 21 - 22 | 3248 | 62 | ||
| AK122202.1 | - | 21 - 22 | 3236 | 62 | ||
| AK147336.1 | - | 20 - 21 | 5840 | 62 | ||
| AK147495.1 | - | 20 - 21 | 5673 | 62 | ||
| AK164654.1 | - | 2 - 3 | 169 | 62 | ||
| BC011533.1 | - | 21 - 22 | 3234 | 62 | ||
| BC054837.1 | - | 20 - 21 | 3039 | 6490394 | 62 | |
| BC057198.1 | - | 22 - 23 | 3386 | 5310414 | 62 | |
| DQ913734.1 | - | 20 - 21 | 2902 | 62 | ||
More Information
Set Membership: |
|

Mbtps1
Genomic Map