Assay Details
| Gene Symbol: Mbtps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167910.1 | NP_001161382.1 | 8 - 9 | 1471 | 79 | |
| NM_019709.4 | NP_062683.3 | 9 - 10 | 1620 | 79 | ||
| GenBank mRNA | AF094820.1 | - | 7 - 8 | 1039 | 79 | |
| AK029048.1 | - | 8 - 9 | 1448 | 79 | ||
| AK122202.1 | - | 8 - 9 | 1437 | 79 | ||
| AK147336.1 | - | 7 - 8 | 4040 | 79 | ||
| AK147495.1 | - | 8 - 9 | 1445 | 79 | ||
| AK209637.1 | - | 2 - 3 | 93 | 79 | ||
| BC011533.1 | - | 8 - 9 | 1437 | 79 | ||
| BC054837.1 | - | 7 - 8 | 1239 | 6490394 | 79 | |
| BC057198.1 | - | 9 - 10 | 1586 | 5310414 | 79 | |
| DQ913734.1 | - | 7 - 8 | 1102 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mbtps1
Genomic Map