| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100018748.mm.1 |
NM_001195088.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018748.mm.1 |
NM_001195088.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018748.mm.1 |
NM_001195089.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018748.mm.1 |
NM_001195089.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018748.mm.1 |
NM_001195090.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018748.mm.1 |
NM_001195090.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018748.mm.1 |
NM_181856.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018748.mm.1 |
NM_181856.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018748.mm.1 |
TC1100001969.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100018752.mm.1 |
NM_001195088.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018752.mm.1 |
NM_001195088.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018752.mm.1 |
NM_001195089.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018752.mm.1 |
NM_001195089.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018752.mm.1 |
NM_001195090.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018752.mm.1 |
NM_001195090.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018752.mm.1 |
NM_181856.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018752.mm.1 |
NM_181856.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018752.mm.1 |
TC1100001969.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100018749.mm.1 |
NM_001195088.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018749.mm.1 |
NM_001195088.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018749.mm.1 |
NM_001195089.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018749.mm.1 |
NM_001195089.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018749.mm.1 |
NM_001195090.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018749.mm.1 |
NM_001195090.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018749.mm.1 |
NM_181856.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018749.mm.1 |
NM_181856.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018749.mm.1 |
TC1100001969.mm.1 |
MTA TC |