Assay Details
| Gene Symbol: Abca8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153145.4 | NP_694785.3 | 12 - 13 | 1690 | 99 | |
| GenBank mRNA | AB255881.1 | - | 11 - 12 | 1609 | 99 | |
| AB255882.1 | - | 12 - 13 | 1697 | 99 | ||
| AF498360.1 | - | 11 - 12 | 1618 | 99 | ||
| AY732492.1 | - | 11 - 12 | 1618 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abca8a
Genomic Map