Assay Details
| Gene Symbol: Abca8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153145.4 | NP_694785.3 | 13 - 14 | 1860 | 98 | |
| GenBank mRNA | AB255881.1 | - | 12 - 13 | 1779 | 98 | |
| AB255882.1 | - | 13 - 14 | 1867 | 98 | ||
| AF498360.1 | - | 12 - 13 | 1788 | 98 | ||
| AY732492.1 | - | 12 - 13 | 1788 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abca8a
Genomic Map