Assay Details
| Gene Symbol: Abca8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153145.4 | NP_694785.3 | 21 - 22 | 2963 | 78 | |
| GenBank mRNA | AB255881.1 | - | 20 - 21 | 2882 | 78 | |
| AB255882.1 | - | 21 - 22 | 2970 | 78 | ||
| AF498360.1 | - | 20 - 21 | 2891 | 78 | ||
| AK031843.1 | - | 2 - 3 | 194 | 78 | ||
| AK148846.1 | - | 1 - 2 | 53 | 78 | ||
| AY732492.1 | - | 20 - 21 | 2891 | 78 | ||
| BC026496.1 | - | 1 - 2 | 88 | 78 | ||
| BC060032.1 | - | 3 - 4 | 457 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abca8a
Genomic Map