Assay Details
| Gene Symbol: Abca8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153145.4 | NP_694785.3 | 23 - 24 | 3214 | 88 | |
| GenBank mRNA | AB255881.1 | - | 22 - 23 | 3133 | 88 | |
| AB255882.1 | - | 23 - 24 | 3221 | 88 | ||
| AF498360.1 | - | 22 - 23 | 3142 | 88 | ||
| AK031843.1 | - | 4 - 5 | 445 | 88 | ||
| AY732492.1 | - | 22 - 23 | 3142 | 88 | ||
| BC026496.1 | - | 3 - 4 | 339 | 88 | ||
| BC060032.1 | - | 5 - 6 | 708 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abca8a
Genomic Map