Assay Details
| Gene Symbol: Hck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172117.1 | NP_001165588.1 | 3 - 4 | 445 | 74 | |
| NM_010407.4 | NP_034537.2 | 3 - 4 | 445 | 74 | ||
| GenBank mRNA | AK149736.1 | - | 3 - 4 | 411 | 74 | |
| AK150290.1 | - | 3 - 4 | 419 | 74 | ||
| AK150709.1 | - | 3 - 4 | 428 | 74 | ||
| AK152506.1 | - | 3 - 4 | 419 | 74 | ||
| AK155975.1 | - | 3 - 4 | 358 | 74 | ||
| AK159234.1 | - | 3 - 4 | 377 | 74 | ||
| AK165315.1 | - | 3 - 4 | 410 | 74 | ||
| BC010478.1 | - | 3 - 4 | 441 | 3978518 | 74 | |
| J03023.1 | - | 3 - 4 | 334 | 74 | ||
| Y00487.1 | - | 3 - 4 | 340 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hck
Genomic Map