Assay Details
| Gene Symbol: Sec14l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166506.1 | NP_001159978.1 | 14 - 15 | 2076 | 105 | |
| NM_001166507.1 | NP_001159979.1 | 14 - 15 | 2076 | 105 | ||
| NM_028777.3 | NP_083053.2 | 14 - 15 | 2076 | 105 | ||
| GenBank mRNA | AK004818.1 | - | 14 - 15 | 2001 | 105 | |
| AK172947.1 | - | 11 - 12 | 1611 | 105 | ||
| AK196113.1 | - | 152 | 105 | |||
| BC005766.1 | - | 14 - 15 | 2034 | 2649054 | 105 | |
| BC086620.1 | - | 14 - 15 | 2009 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sec14l1
Genomic Map