Assay Details
| Gene Symbol: Pcif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146129.3 | NP_666241.1 | 12 - 13 | 1647 | 57 | |
| GenBank mRNA | AK089358.1 | - | 12 - 13 | 1648 | 57 | |
| BC031431.1 | - | 12 - 13 | 1616 | 4241283 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pcif1
Genomic Map