Assay Details
| Gene Symbol: Cdh22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174988.3 | NP_778153.2 | 1 - 2 | 182 | 143 | |
| GenBank mRNA | AB019618.1 | - | 1 - 2 | 172 | 143 | |
| AK140248.1 | - | 1 - 2 | 299 | 143 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB019618.1, NM_174988.3 transcripts.
Set Membership: |
|

Cdh22
Genomic Map