Assay Details
| Gene Symbol: Zfp827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001294279.1 | NP_001281208.1 | 3 - 4 | 1505 | 58 | |
| NM_178267.3 | NP_839998.2 | 3 - 4 | 1505 | 58 | ||
| GenBank mRNA | AK154905.1 | - | 3 - 4 | 1358 | 58 | |
| BC151183.1 | - | 3 - 4 | 1390 | 9088072 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp827
Genomic Map