Assay Details
| Gene Symbol: Zfp827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001294279.1 | NP_001281208.1 | 4 - 5 | 1991 | 85 | |
| NM_178267.3 | NP_839998.2 | 4 - 5 | 1991 | 85 | ||
| GenBank mRNA | AK154905.1 | - | 4 - 5 | 1844 | 85 | |
| BC151183.1 | - | 4 - 5 | 1876 | 9088072 | 85 | |
| CN461771.1 | - | 1 - 2 | 445 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp827
Genomic Map