Assay Details
| Gene Symbol: Gaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159324.1 | NP_001152796.1 | 17 - 18 | 2657 | 83 | |
| NM_008064.3 | NP_032090.3 | 17 - 18 | 2747 | 83 | ||
| GenBank mRNA | AK052211.1 | - | 17 - 18 | 2728 | 83 | |
| AK088481.1 | - | 17 - 18 | 2723 | 83 | ||
| AK139333.1 | - | 17 - 18 | 2695 | 83 | ||
| AK141546.1 | - | 17 - 18 | 2730 | 83 | ||
| AK145631.1 | - | 16 - 17 | 2163 | 83 | ||
| AK146538.1 | - | 17 - 18 | 2651 | 83 | ||
| AK150970.1 | - | 17 - 18 | 2705 | 83 | ||
| BC010210.1 | - | 17 - 18 | 2711 | 3969127 | 83 | |
| U49351.1 | - | 17 - 18 | 2599 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gaa
Genomic Map