Assay Details
| Gene Symbol: Gaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159324.1 | NP_001152796.1 | 3 - 4 | 858 | 81 | |
| NM_008064.3 | NP_032090.3 | 3 - 4 | 948 | 81 | ||
| GenBank mRNA | AK052211.1 | - | 3 - 4 | 929 | 81 | |
| AK088481.1 | - | 3 - 4 | 924 | 81 | ||
| AK139333.1 | - | 3 - 4 | 896 | 81 | ||
| AK141546.1 | - | 3 - 4 | 931 | 81 | ||
| AK145631.1 | - | 2 - 3 | 363 | 81 | ||
| AK146538.1 | - | 3 - 4 | 852 | 81 | ||
| AK150970.1 | - | 3 - 4 | 906 | 81 | ||
| BC010210.1 | - | 3 - 4 | 912 | 3969127 | 81 | |
| U49351.1 | - | 3 - 4 | 800 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gaa
Genomic Map