Assay Details
| Gene Symbol: Gaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159324.1 | NP_001152796.1 | 6 - 7 | 1246 | 64 | |
| NM_008064.3 | NP_032090.3 | 6 - 7 | 1336 | 64 | ||
| GenBank mRNA | AK052211.1 | - | 6 - 7 | 1317 | 64 | |
| AK088481.1 | - | 6 - 7 | 1312 | 64 | ||
| AK139333.1 | - | 6 - 7 | 1284 | 64 | ||
| AK140042.1 | - | 6 - 7 | 1303 | 64 | ||
| AK141546.1 | - | 6 - 7 | 1319 | 64 | ||
| AK145631.1 | - | 5 - 6 | 751 | 64 | ||
| AK146538.1 | - | 6 - 7 | 1240 | 64 | ||
| AK150970.1 | - | 6 - 7 | 1294 | 64 | ||
| BC010210.1 | - | 6 - 7 | 1300 | 3969127 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gaa
Genomic Map