| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200042330.mm.1 |
NM_018888.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042330.mm.1 |
NM_018888.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042330.mm.1 |
NR_024487.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042330.mm.1 |
NR_024487.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042330.mm.1 |
TC0200004975.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200042313.mm.1 |
NM_018888.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042313.mm.1 |
NM_018888.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042313.mm.1 |
NR_024487.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042313.mm.1 |
NR_024487.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042313.mm.1 |
TC0200004975.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200042306.mm.1 |
NM_018888.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042306.mm.1 |
NM_018888.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042306.mm.1 |
NR_024487.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042306.mm.1 |
NR_024487.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042306.mm.1 |
TC0200004975.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200042327.mm.1 |
NM_018888.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042327.mm.1 |
NM_018888.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042327.mm.1 |
NR_024487.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042327.mm.1 |
NR_024487.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042327.mm.1 |
TC0200004975.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200042314.mm.1 |
NM_018888.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042314.mm.1 |
NM_018888.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042314.mm.1 |
NR_024487.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042314.mm.1 |
NR_024487.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042314.mm.1 |
TC0200004975.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200042315.mm.1 |
NM_018888.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042315.mm.1 |
NM_018888.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042315.mm.1 |
NR_024487.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042315.mm.1 |
NR_024487.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042315.mm.1 |
TC0200004975.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200042328.mm.1 |
NM_018888.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042328.mm.1 |
NM_018888.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042328.mm.1 |
NR_024487.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042328.mm.1 |
NR_024487.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042328.mm.1 |
TC0200004975.mm.1 |
MTA TC |