Assay Details
| Gene Symbol: Polg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015810.2 | NP_056625.2 | 2 - 3 | 705 | 84 | |
| NR_027785.1 | 2 - 3 | 558 | 84 | |||
| GenBank mRNA | AF177202.1 | - | 2 - 3 | 610 | 84 | |
| AK041591.1 | - | 2 - 3 | 677 | 84 | ||
| AK042777.1 | - | 2 - 3 | 516 | 84 | ||
| AK164730.1 | - | 2 - 3 | 563 | 84 | ||
| AK169977.1 | - | 2 - 3 | 947 | 84 | ||
| BC119131.1 | - | 2 - 3 | 634 | 8733880 | 84 | |
| BC144810.1 | - | 2 - 3 | 509 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Polg2
Genomic Map