Assay Details
| Gene Symbol: Ern1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023913.2 | NP_076402.1 | 16 - 17 | 2176 | 69 | |
| GenBank mRNA | AB031332.1 | - | 16 - 17 | 2176 | 69 | |
| AK077041.1 | - | 2199 | 69 | |||
| AK080709.1 | - | 10 - 11 | 1433 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ern1
Genomic Map