Assay Details
| Gene Symbol: Cep131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009734.3 | NP_033864.3 | 14 - 15 | 1902 | 88 | |
| GenBank mRNA | AK133245.1 | - | 14 - 15 | 1904 | 88 | |
| AK134381.1 | - | 7 - 8 | 835 | 88 | ||
| BC141108.1 | - | 14 - 15 | 1842 | 9055720 | 88 | |
| D43921.1 | - | 14 - 15 | 1897 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cep131
Genomic Map