Assay Details
| Gene Symbol: Cep131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009734.3 | NP_033864.3 | 17 - 18 | 2252 | 95 | |
| GenBank mRNA | AK133245.1 | - | 17 - 18 | 2255 | 95 | |
| AK134381.1 | - | 10 - 11 | 1185 | 95 | ||
| BC141108.1 | - | 17 - 18 | 2192 | 9055720 | 95 | |
| D43921.1 | - | 17 - 18 | 2247 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cep131
Genomic Map