Assay Details
| Gene Symbol: Dram1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027878.2 | NP_082154.2 | 1 - 2 | 288 | 102 | |
| GenBank mRNA | AK004552.1 | - | 1 - 2 | 288 | 102 | |
| AK151184.1 | - | 1 - 2 | 214 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dram1
Genomic Map