Assay Details
| Gene Symbol: Gnpat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010322.3 | NP_034452.3 | 2 - 3 | 459 | 117 | |
| GenBank mRNA | AF110769.1 | - | 2 - 3 | 423 | 117 | |
| AJ132012.1 | - | 2 - 3 | 254 | 117 | ||
| AK004630.1 | - | 2 - 3 | 433 | 117 | ||
| AK004771.1 | - | 2 - 3 | 439 | 117 | ||
| AK010896.1 | - | 2 - 3 | 433 | 117 | ||
| AK150810.1 | - | 2 - 3 | 417 | 117 | ||
| AK166997.1 | - | 2 - 3 | 431 | 117 | ||
| BC003225.1 | - | 2 - 3 | 431 | 117 | ||
| BC025972.1 | - | 2 - 3 | 416 | 4020505 | 117 | |
| BI990299.1 | - | 2 - 3 | 459 | 117 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gnpat
Genomic Map