Assay Details
| Gene Symbol: Gnpat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010322.3 | NP_034452.3 | 5 - 6 | 897 | 121 | |
| GenBank mRNA | AF110769.1 | - | 5 - 6 | 861 | 121 | |
| AJ132012.1 | - | 5 - 6 | 692 | 121 | ||
| AK004630.1 | - | 5 - 6 | 871 | 121 | ||
| AK004771.1 | - | 5 - 6 | 877 | 121 | ||
| AK010896.1 | - | 5 - 6 | 871 | 121 | ||
| AK150810.1 | - | 5 - 6 | 855 | 121 | ||
| AK166997.1 | - | 5 - 6 | 869 | 121 | ||
| BC003225.1 | - | 5 - 6 | 839 | 121 | ||
| BC025972.1 | - | 5 - 6 | 854 | 4020505 | 121 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gnpat
Genomic Map